Digestion Tubes, Quickfit®
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
Designed for use with block heaters. Ground socket to accept air condensers.
Product Information
- Catalog #Supplier #CapacityLengthExt.ØSocket sizePkAvailabilityPrice
- Supplier Article Number: TJ15/16/1Local Article Number: BIBBTJ15/16/1Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: TJ75/26/3Local Article Number: BIBBTJ75/26/3Specifications:
Specifications
Environmentally Preferable - BIBBTJ100/26/3TJ100/26/3100 ml300 mm26 mm24/29 NS20Log in for availability1 * 20 Itemskr 12 700,00Supplier Article Number: TJ100/26/3Local Article Number: BIBBTJ100/26/3Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: TJ250/42/6Local Article Number: BIBBTJ250/42/6Specifications:
Specifications
Environmentally Preferable