Anti-Phosphothreonine Mouse Monoclonal Antibody [clone: 18F6]
Catalog # CAYM10221-100
Supplier: Cayman Chemical
- Antibody type:Primary
- Antigen name:Phosphothreonine
- Antigen symbol:pT
- Clonality:Monoclonal
- Clone:18F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Storage buffer:containing 0,1% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Size:100 µl
- Pk:100 µl
About this item
Type: Primary
Antigen: Phosphothreonine
Clonality: Monoclonal
Clone: 18F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: