Anti-ST6GAL1 Rabbit Polyclonal Antibody

Leverandør: Abgent
ABGEAP19891CEA 4025 NOK
ABGEAP19891C
Anti-ST6GAL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ST6GAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR