Peqlab peqGOLD Cycle-Pure™, Agarose Gel Extraction Kit
Leverandør: Avantor
K220-50RXNEA 6075 NOK
VWRCK220-50RXN
Peqlab peqGOLD Cycle-Pure™, Agarose Gel Extraction Kit
Agarose gel extraction kit is a fast, spin column-based kit for the quick isolation of DNA fragments (40 bp to 100 kb) from agarose slices.
  • DNA isolation in 10 minutes
  • TAE/TBE agarose gel compatible
  • Spin column format
In just 10 minutes, ultra-pure DNA can be extracted that is ready to use in restriction enzyme digestion, labelling, ligation, transformation, in vitro transcription and sequencing protocols. All reagents are room temperature stable.
Leveringsinformasjon: Kit includes Binding Buffer, 40 ml; Wash Buffer, 50 ml; spin columns, 50; and collection tubes, 50. Contains sufficient material for 50 isolations.