Spill kits, ADR
: Brady
ADR spill kits are eco-friendly and portable and come in a durable see-through bag with shoulder strap. The small ADR chemical kit is designed for small spills of chemical and hazardous liquid. The large ADR maintenance kit is designed for small spills of industrial oil, water, solvents and coolants. The Re-Form ADR kit S is designed for small spills of industrial liquids except for highly aggressive acids and caustics.
- Quick and easy access to the sorbents
- Durable see-through bag makes it easy to check the spill response supplies
- Pk
- Small ADR chemical spill kit, 37 litres, includes 25 pads (41×51 mm), 4 socs (76×1220 mm), 1 pair gloves and 1 disposal bagSmall ADR chemical spill kit, 37 litres, includes 25 pads (41×51 mm), 4 socs (76×1220 mm), 1 pair gloves and 1 disposal bag121-01528138641 KIT1 * 1 Eachkr 1 750,00: 813864: BRDY813864
Specifications
Environmentally Preferable - Large ADR chemical spill kit, 57 litres, includes 35 pads (41×51 mm), 1 socs (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (230×230 mm), 2 yellow disposal
bags and 1 pair of glovesLarge ADR chemical spill kit, 57 litres, includes 35 pads (41×51 mm), 1 socs (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (230×230 mm), 2 yellow disposal
bags and 1 pair of gloves121-01938138671 KIT1 * 1 Eachkr 2 325,00: 813867: BRDY813867Specifications
Environmentally Preferable - Large ADR maintenance spill kit, 63 litres, includes 35 pads (410×510 mm), 1 soc (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (250×250 mm), 1 pair gloves and 2 disposal bagsLarge ADR maintenance spill kit, 63 litres, includes 35 pads (410×510 mm), 1 soc (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (250×250 mm), 1 pair gloves and 2 disposal bags121-01538138661 KIT1 * 1 Eachkr 2 250,00: 813866: BRDY813866
Specifications
Environmentally Preferable - Re-Form ADR maintenance spill kit S, includes 25 pads (410×510 mm), 4 soc (76×1220 mm), 1 pair gloves and 1 disposal bagsRe-Form ADR maintenance spill kit S, includes 25 pads (410×510 mm), 4 soc (76×1220 mm), 1 pair gloves and 1 disposal bags121-01548139551 KIT1 * 1 Eachkr 1 600,00: 813955: BRDY813955
Specifications
Environmentally Preferable