Heat sealing foil, ClearASeal Peel™
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The ClearASeal Peel is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates
- Moderate solvent resistance, can be utilized for short term compound storage at room temperature
- Available as sheets, for use with manual and semi-automated sealers, or in multiple roll formats compatible with specified automated heat sealers
- Sheet seals are inter-leaved with paper sheets to help denote which side is the sealing side and aid removal of one sheet at a time from the pack
The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
The ClearASeal Peel forms a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).
- Optical propertySterilePk
- LabRoll™, 500 m × 78 mmLabRoll™, 500 m × 78 mm731-0475IST-101-078LROptically clear-1 Roll1 * 1 Rullkr 7 300,00: IST-101-078LR: ISTSIST-101-078LR
Specifications
Environmentally Preferable - LabRoll™, 500 m × 78 mmLabRoll™, 500 m × 78 mm731-0476IST-101-078SROptically clear+1 Roll1 * 1 Rullkr 10 150,00: IST-101-078SR: ISTSIST-101-078SR
Specifications
Environmentally Preferable - LabRoll™, trial pack, 5 m × 78 mmLabRoll™, trial pack, 5 m × 78 mm731-0481IST-101-078TROptically clear-1 Roll1 * 1 Rullkr 700,00: IST-101-078TR: ISTSIST-101-078TR
Specifications
Environmentally Preferable - LabRoll™, 350 m × 115 mmLabRoll™, 350 m × 115 mm731-0477IST-101-115LROptically clear-1 Roll1 * 1 Rullkr 6 550,00: IST-101-115LR: ISTSIST-101-115LR
Specifications
Environmentally Preferable - LabRoll™, 350 m × 115 mmLabRoll™, 350 m × 115 mm731-0478IST-101-115SROptically clear+1 Roll1 * 1 Rullkr 8 650,00: IST-101-115SR: ISTSIST-101-115SR
Specifications
Environmentally Preferable - LabRoll™, trial pack, 5 m × 115 mmLabRoll™, trial pack, 5 m × 115 mm731-0482IST-101-115TROptically clear-1 Roll1 * 1 Rullkr 700,00: IST-101-115TR: ISTSIST-101-115TR
Specifications
Environmentally Preferable - LabSheet™, 125 m × 78 mmLabSheet™, 125 m × 78 mm731-0479IST-101-078LSOptically clear-1001 * 100 Itemskr 740,00: IST-101-078LS: ISTSIST-101-078LS
Specifications
Environmentally Preferable - LabSheet™, 125 m × 78 mmLabSheet™, 125 m × 78 mm731-0480IST-101-078SSOptically clear+1001 * 100 Itemskr 1 350,00: IST-101-078SS: ISTSIST-101-078SS
Specifications
Environmentally Preferable - LabSheet™, trial pack, 125×78 mmLabSheet™, trial pack, 125×78 mm731-0483IST-101-078TSOptically clear-51 * 5 Itemskr 240,00: IST-101-078TS: ISTSIST-101-078TS
Specifications
Environmentally Preferable