These two portable meters are designed for different applications: CO 300 is capable of measuring conductivity, salinity, TDS and temperature values and can be universally used. Typical applications include: Fresh and sea water aquaristics, fish farming, water and drinking water monitoring. LCO 302 measures conductivity, resistivity and temperature and is specifically designed for ultra-pure water applications. It can also be used for functional checks of ion exchangers.
- Made of impact resistant ABS, front side is IP 65 rated
- Min./Max. value memory, 'HOLD' function, auto-off function
- Automatic temperature compensation via integrated temperature sensor
- Supplied ready-to-use with measuring cells
Leveringsinformasjon: CO 300 is supplied with 2-pole measuring cell (Ø 12 mm, graphite) with 1,2 m cable, battery, calibration protocol and manual. LCO 302 is supplied with 2-pole measuring cell (Ø 12 mm, stainless steel) with 1,2 m cable, battery and manual.