Drape sheets, Foliodrape® Comfort
Leverandør: Hartmann
Foliodrape® Comfort
HART2775472EA 1125 NOK
HART2775472
Drape sheets, Foliodrape® Comfort
Førstehjelpsutstyr Kirurgiske overtrekk
These surgical drape sheets are made from a soft three layer material. For sterile draping of patients or equipment in the operating theatre.
  • Impermeable to moisture and bacteria
  • Abrasion resistant, extremely low linting and highly conformable