VWR®, Test Sieves, 300×30 mm, with Square Holes
- ØHøydeMaskeviddeFp
- : VWRI510-2678
Specifications
Environmentally Preferable - : VWRI510-2658
Specifications
Environmentally Preferable - : VWRI510-2680
Specifications
Environmentally Preferable - : VWRI510-2679
Specifications
Environmentally Preferable - : VWRI510-2660
Specifications
Environmentally Preferable - : VWRI510-2682
Specifications
Environmentally Preferable - : VWRI510-2681
Specifications
Environmentally Preferable - : VWRI510-2659
Specifications
Environmentally Preferable - : VWRI510-2684
Specifications
Environmentally Preferable - : VWRI510-2662
Specifications
Environmentally Preferable - : VWRI510-2683
Specifications
Environmentally Preferable - : VWRI510-2661
Specifications
Environmentally Preferable - : VWRI510-2664
Specifications
Environmentally Preferable - 510-2686300 mm30 mm100,00 mm11 * 1 Itemskr 3 475,00: VWRI510-2686
Specifications
Environmentally Preferable - : VWRI510-2663
Specifications
Environmentally Preferable - : VWRI510-2685
Specifications
Environmentally Preferable - : VWRI510-2666
Specifications
Environmentally Preferable - 510-2687300 mm30 mm106,00 mm11 * 1 Itemskr 3 475,00: VWRI510-2687
Specifications
Environmentally Preferable - : VWRI510-2665
Specifications
Environmentally Preferable - : VWRI510-2667
Specifications
Environmentally Preferable - 510-2689300 mm30 mm125,00 mm11 * 1 Itemskr 3 475,00: VWRI510-2689
Specifications
Environmentally Preferable - 510-2688300 mm30 mm112,00 mm11 * 1 Itemskr 3 475,00: VWRI510-2688
Specifications
Environmentally Preferable - : VWRI510-2669
Specifications
Environmentally Preferable - : VWRI510-2668
Specifications
Environmentally Preferable - : VWRI510-2671
Specifications
Environmentally Preferable - : VWRI510-2649
Specifications
Environmentally Preferable - : VWRI510-2670
Specifications
Environmentally Preferable - : VWRI510-2673
Specifications
Environmentally Preferable - : VWRI510-2651
Specifications
Environmentally Preferable - : VWRI510-2672
Specifications
Environmentally Preferable - : VWRI510-2650
Specifications
Environmentally Preferable - : VWRI510-2653
Specifications
Environmentally Preferable - : VWRI510-2675
Specifications
Environmentally Preferable - : VWRI510-2674
Specifications
Environmentally Preferable - : VWRI510-2652
Specifications
Environmentally Preferable - : VWRI510-2655
Specifications
Environmentally Preferable - : VWRI510-2677
Specifications
Environmentally Preferable - : VWRI510-2676
Specifications
Environmentally Preferable - : VWRI510-2654
Specifications
Environmentally Preferable - : VWRI510-2657
Specifications
Environmentally Preferable - : VWRI510-2656
Specifications
Environmentally Preferable