HPLC columns, LiChroCART®, Superspher® 60 Si
Supplier: Merck
Certificates
About this item
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- 1.50201.71421.50201.7142AnalyticalSi60 Å4 µm2,00 mm10 mmLog in for availability1 * 3 Itemskr 2 860,00Specifications:
Specifications
Environmentally Preferable - 1.50229.71421.50229.7142AnalyticalSi60 Å4 µm2,00 mm30 mmLog in for availability1 * 1 Itemskr 4 540,00Specifications:
Specifications
Environmentally Preferable - 1.50234.71421.50234.7142AnalyticalSi60 Å4 µm2,00 mm55 mmLog in for availability1 * 1 Itemskr 6 000,00Specifications:
Specifications
Environmentally Preferable - Specifications:
Specifications
Environmentally Preferable - 1.50233.71421.50233.7142AnalyticalSi60 Å4 µm3,00 mm30 mmLog in for availability1 * 1 Itemskr 4 540,00Specifications:
Specifications
Environmentally Preferable - 1.50236.71421.50236.7142AnalyticalSi60 Å4 µm3,00 mm55 mmLog in for availability1 * 1 Itemskr 5 600,00Specifications:
Specifications
Environmentally Preferable - 1.50175.71421.50175.7142AnalyticalSi60 Å4 µm3,00 mm125 mmLog in for availability1 * 1 Itemskr 9 125,00Specifications:
Specifications
Environmentally Preferable - 1.50177.71421.50177.7142AnalyticalSi60 Å4 µm3,00 mm250 mmLog in for availability1 * 1 Itemskr 10 725,00Specifications:
Specifications
Environmentally Preferable - Specifications:
Specifications
Environmentally Preferable - 1.50302.71421.50302.7142AnalyticalSi60 Å4 µm4,00 mm30 mmLog in for availability1 * 1 Itemskr 4 225,00Specifications:
Specifications
Environmentally Preferable - 1.50228.71421.50228.7142AnalyticalSi60 Å4 µm4,00 mm55 mmLog in for availability1 * 1 Itemskr 5 600,00Specifications:
Specifications
Environmentally Preferable - 1.50171.71421.50171.7142AnalyticalSi60 Å4 µm4,00 mm75 mmLog in for availability1 * 1 Itemskr 7 500,00Specifications:
Specifications
Environmentally Preferable - 1.50170.71421.50170.7142AnalyticalSi60 Å4 µm4,00 mm125 mmLog in for availability1 * 1 Itemskr 9 125,00Specifications:
Specifications
Environmentally Preferable - 1.50174.71421.50174.7142AnalyticalSi60 Å4 µm4,00 mm250 mmLog in for availability1 * 1 Itemskr 10 725,00Specifications:
Specifications
Environmentally Preferable - 1.51448.71421.51448.7142AnalyticalSi60 Å4 µm4,60 mm100 mmLog in for availability1 * 1 Itemskr 9 125,00Specifications:
Specifications
Environmentally Preferable - 1.51442.71421.51442.7142AnalyticalSi60 Å4 µm4,60 mm125 mmLog in for availability1 * 1 Itemskr 9 125,00Specifications:
Specifications
Environmentally Preferable - 1.51432.71421.51432.7142AnalyticalSi60 Å4 µm4,60 mm150 mmLog in for availability1 * 1 Itemskr 9 400,00Specifications:
Specifications
Environmentally Preferable - 1.51431.71421.51431.7142AnalyticalSi60 Å4 µm4,60 mm250 mmLog in for availability1 * 1 Itemskr 10 725,00Specifications:
Specifications
Environmentally Preferable - 1.50178.71421.50178.7142Semi-preparativeSi60 Å4 µm10,00 mm10 mmLog in for availability1 * 2 Itemskr 4 980,00Specifications:
Specifications
Environmentally Preferable - 1.51443.71421.51443.7142Semi-preparativeSi60 Å4 µm10,00 mm125 mmLog in for availability1 * 1 Itemskr 19 740,00Specifications:
Specifications
Environmentally Preferable - 1.51444.71421.51444.7142Semi-preparativeSi60 Å4 µm10,00 mm150 mmLog in for availability1 * 1 Itemskr 20 620,00Specifications:
Specifications
Environmentally Preferable - 1.50179.71421.50179.7142Semi-preparativeSi60 Å4 µm10,00 mm250 mmLog in for availability1 * 1 Itemskr 24 525,00Specifications:
Specifications
Environmentally Preferable