Digital rotational viscometers, evo expert
Leverandør: Fungilab
FUNGVE320003EA 32000 NOK
FUNGVE320003 FUNGVE320002 FUNGVE321003 FUNGVE321002 FUNGVE320001 FUNGVE321001
Digital rotational viscometers, evo expert
The evo expert viscosimeters are designed for advanced performance and multifunctional measurements.
  • Colour TFT screen enabling easy readings as well as fast data recognition
  • Intuitive sensitive keypad to select your data in one touch with light-up LED
  • USB 2.0 connection for downloading data to your computer
  • Integrated Wi-Fi system for easy control and access to your measurements
  • Viscosity reading include dynamic viscosity (cP or mPa·s) or kinematics viscosity (cSt)
(L) Low to medium viscosity - (R) Medium to high viscosity - (H) High to very high viscosity