Norway
Order Entry
Biohazard Waste Containers
Biohazard Waste Containers
Certificates
undefined
Biohazard Waste Containers
Product Information
  • Catalog #
    Supplier #
    Colour
    Capacity
    Ø×H
    Pk
    Availability
    Price
  • JUSU05930WH
    05930WH
    White
    34 L
    354×464 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 700,00
    kr 2 700,00
    1 * 1 Items
       kr 2 700,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • JUSU05930RD
    05930RD
    Red
    34 L
    354×464 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 850,00
    kr 1 850,00
    1 * 1 Items
       kr 1 850,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable