C300 wet Cut Protection Gloves, 60542 Series
Supplier: UVEX
Certificates
About this item
The C300 cut protection glove range sets standards for protection, comfort, tactility, and efficiency. This high-tech safety glove combines all of these qualities with level 3/C cut protection.
- Highly flexible
- Very good tactile sensation
- Perfect fit with 3D ergo technology
- Outstanding wearer comfort due to uvex climazone
A cut protection glove with excellent wearer comfort. Palm and fingertips feature a High-Performance Elastomer (HPE) coating. Free of harmful substances in accordance with OEKO-TEX® standard 100.
Specifications
- Catalog No:
- 112-4431
- 112-4430
- 112-4433
- 112-4432
- 112-4429
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourPkAvailabilityPrice
- Supplier Article Number: 6054207Local Article Number: UVEX6054207Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018 - Supplier Article Number: 6054208Local Article Number: UVEX6054208Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018 - Supplier Article Number: 6054209Local Article Number: UVEX6054209Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018 - Supplier Article Number: 6054210Local Article Number: UVEX6054210Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018 - Supplier Article Number: 6054211Local Article Number: UVEX6054211Specifications:
More
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableDisposablePowdered / powder-freePowder-freeEN normEN ISO 21420:2020/EN 388:2016 + A1:2018