Stopcocks, straight bore, SP Wilmad-LabGlass
Leverandør: SP Industries
WILMLG-9600-102EA 790 NOK
WILMLG-9600-102 WILMLG-9600-114
Stopcocks, straight bore, SP Wilmad-LabGlass
Glass.
Stopcock plug are ground to interchangeable NS specifications. Plugs are solid and have a 1:10 taper.