Muffelovner, med klappdør eller løftedør, L- og LT-serien
Leverandør: Nabertherm
455-0401EA 59075 NOK
NABEL-091H2TN NABEL-091H1TN NABEL-051K2RN NABEL-051K1RN NABEL-241K2RN NABEL-401H1RN NABEL-241K1RN NABEL-051T3TN NABEL-401H2TN NABEL-401H1TN NABEL-031H1TN NABEL-031H2TN NABEL-151S3RN NABEL-151K2RN NABEL-151K1RN NABEL-091T3TN NABEL-091K1RN NABEL-091K1TN NABEL-013K2EN NABEL-151T3TN NABEL-051H1TN NABEL-091K2RN NABEL-051H2TN NABEL-241H2TN NABEL-031K2RN NABEL-241H1TN NABEL-031K1RN NABEL-401K1RN NABEL-401K2RN NABEL-013K27N NABEL-051S3RN NABEL-091S3RN NABEL-151H1TN NABEL-151H2TN
Muffelovner, med klappdør eller løftedør, L- og LT-serien
Electrically-heated muffle furnaces are ideal for daily laboratory use. Units feature rust-free, textured stainless steel exteriors. The furnaces have Tmax 1100 or 1200 °C. Models feature an adjustable air inlet which is integrated in the door, an exhaust air outlet in the rear wall of the furnace and a silent solid-state power control relay.
  • Heating on two sides by means of ceramic heating plates (heating on three sides for models L 24/11 - LT 40/12)
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to replace
  • Module lining is highly durable cured vacuum fibre
  • Double walled casing for low external temperatures and high stability
  • Fold down door (L models) can be used as work platform or lift door (LT models) with hot surface facing away from the operator
  • Non classified insulation avoids the potential risk of cancer
Leveringsinformasjon: L 1 model supplied with choice of digital R 7 controller with 1 holding temperature or 3216 controller with one program and 8 segments. Other models supplied with choice of Standard B 410 controller with LC display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions or optional C 450 controller with LC display, 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. B 410 and C 450 controllers can automatically activate the vent with fan or catalyst (not models L 15 or LT 15). All units require a 230 V single-phase power supply, except L 24, LT 24 and L 40, LT 40 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of furnace. Manual or automatic gas supply systems are also available.
NB: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
* 400 V, 3 phase power supply.