5-Bromo-3-chloro-2-fluoropyridine 95%
Leverandør: Thermo Scientific
|
Warning
|
H64547.06EA
4975
NOK
H64547.06
5-Bromo-3-chloro-2-fluoropyridine 95%
5-Bromo-3-chloro-2-fluoropyridine
Formel:
C₅H₂BrClFN MW: 210,43 g/mol Smeltepunkt: 60 °C |
MDL nummer:
MFCD09834365 CAS-nummer: 38185-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...