TRIS-glycine-SDS running buffer (10X), pH 8,3
Physical Form: Liquid
Appearance: Colourless
- Physical form:Liquid
- Appearance:Colourless
- pH valuePk
- TRIS-glycine-SDS running buffer (10X)TRIS-glycine-SDS running buffer (10X)J61006.K78.34 L1 * 4 Lkr 2 650,00: J61006.K7: ALFAJ61006.K7
- TRIS-glycine-SDS running buffer (10X)TRIS-glycine-SDS running buffer (10X)J61006.K28.31 L1 * 1 Lkr 955,00: J61006.K2: ALFAJ61006.K2
- TRIS-glycine-SDS running buffer (10X)TRIS-glycine-SDS running buffer (10X)J61006.K38.32 L1 * 2 Lkr 1 600,00: J61006.K3: ALFAJ61006.K3