ALFAJ64257
J64257.MAEA 1625 NOK
J64257.MA
ALFAJ64257
A cell-permeable compound with anti-apoptotic properties.
  • White powder
  • Soluble in water at 5mM or in DMSO at 100mM
Powder
Bax Channel Blocker is a potent inhibitor of Bax-mediated mitochondrial cytochrome C release (IC50 = 0.52 M). It is suggested to inhibit Bax (Bcl-2-associated X protein) channel formation and activity.