/
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
2'-Amino-2'-deoxyguanosine 98%
J64650.03
:
Thermo Fisher Scientific
:
2'-Amino-2'-deoxyguanosine 98%
J64650.03
:
Thermo Fisher Scientific
:
kr 7 725,00
1 * 1 g
-
+
Pk:
1 g
:
60966-26-9
:
:
C₁₀H₁₄N₆O₄
:
MFCD01723954
:
282.26 g/mol
:
GHS07