N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 95%
Leverandør: Thermo Scientific
|
Warning
|
L16739.14EA
1100
NOK
L16739.14
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 95%
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
Formel:
C₇H₁₁F₄NO MW: 201,17 g/mol Kokepunkt: 58 °C (5 mmHg) Tetthet: 1,228 Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00069160 CAS-nummer: 392-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...