Automated plate washing for ELISA, cells and beads
Improve lab efficiency & optimize results with fast and efficient plate washers. Compare and find the optimal plate washer for your applications.
Compare now
TECAN microplate washers
Learn more
Du søkte etter: Buffere, vask
49  treff i ditt søk
SearchResultCount:"49"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Artikkenummer: (J373-4L)
Produsent: VWR Chemicals
Beskrivelse: Washing buffer, PBS buffer solution 10X concentrate (phosphate buffered saline)
UOM: 1 * 4 L
HMS Sertifikater
Artikkenummer: (E715-1L)
Produsent: VWR Chemicals
Beskrivelse: Phosphate Buffered Saline (PBS) with 0,05% Tween® 20, pH 7,5 is a ready-to-use 1× concentration.
UOM: 1 * 1 L
HMS
Artikkenummer: (437117K)
Produsent: VWR Chemicals
Beskrivelse: Bestrålt konsentrat av PBS, egnet for vasking av celler etter fortynning.
UOM: 1 * 10 L
HMS Sertifikater
Artikkenummer: (444037E)
Produsent: VWR Chemicals
Beskrivelse: Steril løsning, pH 7,2, for vasking av blotter.
UOM: 1 * 10 L
HMS Sertifikater
Artikkenummer: (K875-500ML)
Produsent: VWR Chemicals
Beskrivelse: PBS Tween® 20 blot wash buffer is a 20X concentration with 1% Tween® 20.
UOM: 1 * 500 mL
HMS Sertifikater
Produsent: VWR Chemicals
Beskrivelse: Wash buffers ideal for use in applications such as Western blotting and ELISA. Prevents high background by effectively removing unbound reagents.
Artikkenummer: (K859-100TABS)
Produsent: VWR Chemicals
Beskrivelse: Each tablet prepares 100 ml of a 1X TBS solution when dissolved in 100 ml deionised H₂O. 1X TBS contains 25 mM Tris, 140 mM sodium chloride, and ...
UOM: 1 * 1 Each
HMS Sertifikater
Artikkenummer: (444057Y)
Produsent: VWR Chemicals
Beskrivelse: Steril løsning for bruk ved vasking av celler.
UOM: 1 * 10 L
HMS Sertifikater
Artikkenummer: (A9162.0100)
Produsent: PanReac AppliChem
Beskrivelse: Tablets for preparation of 100 ml phosphate buffered saline (PBS).
UOM: 1 * 100 Items
HMS
Produsent: Cayman Chemical
Beskrivelse: 250 mM Tris-base, pH 7,4, 1,37 M sodium chloride, containing 27 mM KCl, 0,4 mM phenol red.
Produsent: G-Biosciences
Beskrivelse: Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.
Artikkenummer: (ROCKMB-013)
Produsent: Rockland Immunochemicals
Beskrivelse: Tween Tris buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be us...
UOM: 1 * 1 L
Produsent: G-Biosciences
Beskrivelse: 10X PBS (Phosphate Buffered Saline) is a 10 fold concentrated, sterile, pH adjusted blend of phosphate buffer and saline solution, which when diluted...
Produsent: Cayman Chemical
Beskrivelse: Formulation: Concentrated TBS (250 mM Tris-base; pH 7,4; 1,37 M sodium chloride and 27 mM KCl).
HMS
Artikkenummer: (786-027)
Produsent: G-Biosciences
Beskrivelse: PBS 10X solution is used as a wash buffers for Western blotting, ELISA and other applications.
UOM: 1 * 500 mL
HMS
Produsent: Rockland Immunochemicals
Beskrivelse: Phosphate buffered and Tris buffered saline are suitable for multiple applications including biological diluent buffer for antibodies or other biologi...
HMS
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 49
no targeter for Bottom