Overhead stirrers, R1
Leverandør: CAT (M. Zipperer GmbH)
CATA60211EA 1875 NOK
CATA60211
Overhead stirrers, R1
Omrørere Omrører
Handy, battery powered mini-stirrer for mixing low viscosity liquids.
  • Ideal for test tubes and other small vessels of up to 500 ml
  • Stirrer requires only one 1,5 V AA battery
  • Simple to use as no cables
  • Choice of 3 stirring rods: Plastic (3 mm Ø)
  • Stainless steel for small vessels or stainless steel with large steel blade for maximum stirring performance