Vacuum pumps, deep vacuum, oil
- Catalog No:
- THSAVLP80DDA-230
- THSAVLP285-230
- Model:
- VLP80DDA-230
- VLP285-230
- Power supply:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Weight:
- 22 kg (shipping weight)
- 22 kg (shipping weight)
- W×D×H:
- 160×420×230 mm, shipping
- 160×420×230 mm, shipping
- Ultimate vacuum:
- 1.95×10³ mbar (1,5 mtorr), max. vacuum
- 1,95×10³ mbar (1,5 mtorr), max. vacuum
- Pk
- Vacuum pump, 283 L/min. displacement at 60 Hz
Accessories
Specifications
ModelVLP285-230Environmentally PreferablePower supply230 V, 50/60 HzWeight22 kg (shipping weight)W×D×H160×420×230 mm, shippingUltimate vacuum1,95×10³ mbar (1,5 mtorr), max. vacuum - High-vacuum pump with VPOF110, 76 L/min. displacement at 60 HzHigh-vacuum pump with VPOF110, 76 L/min. displacement at 60 HzTHSAVLP80DDA-230VLP80DDA-23011 * 1 Items
Accessories
Specifications
ModelVLP80DDA-230Environmentally PreferablePower supply230 V, 50/60 HzWeight22 kg (shipping weight)W×D×H160×420×230 mm, shippingUltimate vacuum1.95×10³ mbar (1,5 mtorr), max. vacuum