Modular heating blocks and accessories
Leverandør: Thermo Fisher Scientific
BARNBK165X20AQEA 2650 NOK
BARNBK165X20AQ
Modular heating blocks and accessories
Termostatblokker Tilbehør til termostatblokker
Constructed from a solid anodised aluminium block.
  • Anodised coating resists the harshest environments
  • Each block has a thermometer well for measuring block temperature
  • Fit all Barnstead/Thermolyne brand modular block dri-baths