Norway
Order Entry
Standards, calibration for LCK cuvettes, Addista
 :  Hach
undefined
Standards, calibration for LCK cuvettes, Addista
 :  Hach
  •  
    Concentration
    For test kit(s)
    Pk
  • LCA 390 Addista Mono standard solution
    LCA 390 Addista Mono standard solution
    662-2360
    LCA390
    AOX: 0,05 - 3,0 mg/l; AOX: 0,005 - 0,5 mg/l
    LCK390, LCK391
    250 mL
    1 * 250 mL
    1 * 250 mL - kr 940,00
    kr 940,00
    1 * 250 mL
       kr 940,00
     : LCA390
     : HACHLCA390
  • LCA 700 Addista Multi parameter standard solution
    LCA 700 Addista Multi parameter standard solution
    662-2362
    LCA700
    Ammonium: 0,015 - 2.0 mg/l NH₄-N; Chloride: 1 - 70 mg/l Cl; Potassium: 5 - 50 mg/l and 8 - 50 mg/l K; Phosphate (ortho): 0,5 - 5,0 mg/l PO₄-P; COD: 5 - 60 mg/l O₂; Total Nitrogen: 5 - 40 mg/l
    LCK304, LCK311, LCK228, LCK 328, LCK348, LCK1414, LCK238
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA700
     : HACHLCA700
  • LCA 701 Addista Multi parameter standard solution
    LCA 701 Addista Multi parameter standard solution
    662-2363
    LCA701
    Lead:, 0,1 - 2,0 mg/l Pb; Iron: 0,2 - 6,0 mg/l Fe; Copper: 0,1 - 8,0 mg/l Cu;
    Nickel: 0,1 - 6,0 mg/l Ni;
    Sulphate: 150 - 900 mg/l SO₄; Zinc: 0,2 - 6,0 mg/l Zn
    LCK306, LCK321, LCK329, LCK337, LCK353, LCK360
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA701
     : HACHLCA701
  • LCA 702 Addista Multi parameter standard solution
    LCA 702 Addista Multi parameter standard solution
    662-2364
    LCA702
    Aluminium: 0,02 - 0,5 mg/l Al; Cadmium: 0,02 - 0,3 mg/l Cd; Chromium ( VI and total) 0,03 - 1,0 mg/l Cr; Chromium (trace): 0,005 - 0,25 mg/l Cr; Sulphate: 150 - 900 mg/l SO₄
    LCK301, LCK308, LCK313, LCK353,
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA702
     : HACHLCA702
  • LCA 703 Addista Multi parameter standard solution
    LCA 703 Addista Multi parameter standard solution
    662-2365
    LCA703
    Orthophosphate: 1,6 - 30mg/l 2 - 20 mg/l PO₄-P; COD: 0 - 1000 mg/l; Ammonium: 2 - 47 mg/l; Chloride: 1 - 70 mg/l; Nitrate 0,23 - 13,5 mg/l; Sulphate TOC
    LCK049, LCK114, LCK303, LCK311, LCK339, LCK350, LCK353, LCK386
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 225,00
    kr 1 225,00
    1 * 1 Each
       kr 1 225,00
     : LCA703
     : HACHLCA703
  • LCA 704 Addista Multi parameter standard solution
    LCA 704 Addista Multi parameter standard solution
    662-2366
    LCA704
    Sulphate: 40 - 150 mg/l SO₄; Ammonium: 1 - 12 mg/l NH₄-N; Chloride: 1 - 70 mg/l Cl; COD: 15 - 150 mg/l; Nitrate: 5 - 35 mg/l NO₃; Phosphate (ortho): 0,05 - 1,5 mg/l PO₄-P; TOC: 3 - 30 mg/l C
    LCK153, LCK305, LCK311, LCK314, LCK340, LCK349, LCK385
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 225,00
    kr 1 225,00
    1 * 1 Each
       kr 1 225,00
     : LCA704
     : HACHLCA704
  • LCA 705 Addista Multi parameter standard solution
    LCA 705 Addista Multi parameter standard solution
    662-2367
    LCA705
    COD:1000 - 10000 mg/l O₂; Ammonium: 47 - 130 mg/l NH₄-N; Chloride, 1 - 70 mg/l C; TOC: 300 - 3000 mg/l C
    LCK014, LCK302, LCK311, LCK387
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA705
     : HACHLCA705
  • LCA 707 Addista Multi parameter standard solution
    LCA 707 Addista Multi parameter standard solution
    662-2369
    LCA707
    Nitrite, 0,015 - 0,6 mg/l NO₂-N; COD: 50 - 300 mg/l O₂; Phosphate (total): 0,5 - 5,0 mg/l PO₄-P
    LCK341, LCK614, LCK348
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 225,00
    kr 1 225,00
    1 * 1 Each
       kr 1 225,00
     : LCA707
     : HACHLCA707
  • LCA 708 Addista Multi parameter standard solution
    LCA 708 Addista Multi parameter standard solution
    662-2370
    LCA708
    LATON 20 - 100 mg/l TNb; COD: 100 - 2000 mg/l O₂; Phosphate (total): 2 - 20 mg/l PO₄-P
    LCK338, LCK514, LCK350
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA708
     : HACHLCA708
  • LCA 310 Addista Mono standard solution
    LCA 310 Addista Mono standard solution
    662-2358
    LCA310
    Chlorine, free: 0,05 - 2,0 mg/l Cl₂ free/ClO₂; Chlorine/Ozone/Chlorine dioxide: 0,05 - 2,0 mg/l Cl₂
    LCK310, LCK410
    1
    1 * 1 Items
    1 * 1 Items - kr 1 050,00
    kr 1 050,00
    1 * 1 Items
       kr 1 050,00
     : LCA310
     : HACHLCA310
  • LCA 333 Addista Mono standard solution
    LCA 333 Addista Mono standard solution
    662-2359
    LCA333
    Surfactants, non-ionic: 0,2 - 6,0 mg/l as TRITON×100
    LCK333
    1
    1 * 1 Items
    1 * 1 Items - kr 990,00
    kr 990,00
    1 * 1 Items
       kr 990,00
     : LCA333
     : HACHLCA333
  • LCA 706 Addista Multi parameter standard solution
    LCA 706 Addista Multi parameter standard solution
    662-2368
    LCA706
    Iron (trace): 0,01 - 1,0 mg/l Fe; Copper (trace): 0,01 - 1,0 mg/l Cu;
    Nickel (trace): 0,05 - 1,0 mg/l Ni; Manganese 0,02 - 5,0 mg/l Mn
    LCK521, LCK529, LCK537, LCW032
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 175,00
    kr 1 175,00
    1 * 1 Each
       kr 1 175,00
     : LCA706
     : HACHLCA706
  • LCA 555 Addista Mono standard solution
    LCA 555 Addista Mono standard solution
    662-2361
    LCA555
    BOD5: 4 - 1650 mg/l O₂
    LCK555
    10 Tests
    1 * 1 Each
    1 * 1 Each - kr 1 025,00
    kr 1 025,00
    1 * 1 Each
       kr 1 025,00
     : LCA555
     : HACHLCA555
  • LCA 709 Addista Multi parameter standard solution
    LCA 709 Addista Multi parameter standard solution
    662-2371
    LCA709
    LATON 1 - 16 mg/l TNb; COD: 50 - 300 mg/l O₂; Phosphate (total): 0,05 - 1,5 mg/l PO₄-P; Nitrite: 0,6 - 6,0 mg/l NO₂-N;
    LCK138, LCK614, LCK349, LCK342
    1 KIT
    1 * 1 Each
    1 * 1 Each - kr 1 225,00
    kr 1 225,00
    1 * 1 Each
       kr 1 225,00
     : LCA709
     : HACHLCA709