VWR® Nitrile White, Nitrile Gloves
Leverandør: VWR
VWR®
112-4513EA 450 NOK
112-4513
VWR® Nitrile White, Nitrile Gloves
Hansker
These thin, ambidextrous gloves are suitable for use in medical applications, food processing and for general applications in non hazardous areas.
  • Non sterile, powder-free, textured fingers
  • Very thin: Provide excellent elasticity for good wearer comfort
  • Free from phthalates/softeners and allergenic latex proteins; particularly skin friendly for users with latex allergy
  • Beaded cuffs for easier donning
  • Low thickness and textured fingertips for excellent grip
Sertifiseringer: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, MDD Class I; PPE Cat. III
Food contact approved.
NB: * Thickness is double measured, minimum values