Norway
Order Entry
Diphenylphosphine 95%, AcroSeal®
Flammable
Irritant
Diphenylphosphine 95%, AcroSeal®
Catalog # 427290500.
CAS Number:  
undefined
Flammable
Irritant
Diphenylphosphine 95%, AcroSeal®
Catalog # 427290500.
CAS Number:  
Supplier Article Number: 427290500
Local Article Number: ACRO427290500
Price
kr 5 025,00
Log in for availability
Unit of Measure 1 * 50 mL

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6250 to 1.6300 (20°C, 589 nm)
  • GC:
    ≥94 %
  • UV:
    at 270 nm A: ≤0.005
  • Residue after evaporation:
    ≤2 ppm
  • Acidity/alkalinity:
    ≤0.0001 meq/g
  • Gradient analysis:
    :
  • Absorbance largest eluted peak:
    ≤0.002 A.U.
  • Filtered to 0.2 micrometer:
    Filtered

Specification Test Results

About this item

 :
 :186.19 g/mol
Density:1.07 g/cm³
Flash Pt:110 °C
Storage Temperature:Ambient
UN:2845
ADR:4.2,I
 :GHS02
 :GHS07
CAS Number:829-85-6
Formula:C₁₂H₁₁P
MDL Number:MFCD00003040