Isovaleric anhydride 95%

Danger

ACRO451090250EA 785 NOK
ACRO451090250 ACRO451091000
Isovaleric anhydride 95%
Isovaleric anhydride
Formel: C₁₀H₁₈O₃
MW: 186,25 g/mol
Kokepunkt: 215 °C
Smeltepunkt: –65 °C
Tetthet: 0,94 g/cm³
Flash punkt: 91 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015051
CAS-nummer: 1468-39-9
FN: 3265
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
Assay ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR