

(1-Methylpiperidin-4-yl)methanamine 97%
ACRO451460100
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
:
:128.22 g/mol
:80…81 °C (20 mmHg)
:Ambient
:2735
:8,II
:GHS07
:GHS05
:7149-42-0
:C₇H₁₆N₂
:MFCD05022430