Syringes, with built-in chaney adapter, Microliter™ and Gastight®
Supplier: HAMILTON BONADUZ
Certificates
About this item
Specifications
- Catalog No:
- HAMI81524
- HAMI81318
- HAMI80505
- HAMI86204
- HAMI81624
- Point style:
- No needle included
- 2
- 2
- 3
- No needle included
- Termination:
- TLL
- LTN
- N
- KH
- TLL
Specifications
Product Information
- Catalog #Supplier #GaugeVolumeLengthTypePkAvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferablePoint style3TerminationKH - Specifications:
More
Specifications
Environmentally PreferablePoint style2TerminationLTN - Specifications:
More
Specifications
Environmentally PreferablePoint style2TerminationN - HAMI8152481524No needle included5 µlNo needle included1005 TLLCH1Log in for availability1 * 1 Itemskr 6 050,00Specifications:
More
Specifications
Environmentally PreferablePoint styleNo needle includedTerminationTLL - HAMI8162481624No needle included10000 µlNo needle included1010 TLLCH1Log in for availability1 * 1 Itemskr 4 000,00Specifications:
More
Specifications
Environmentally PreferablePoint styleNo needle includedTerminationTLL