Norway
Order Entry
Accessories for storage cabinets for acid Protector®
Accessories for storage cabinets for acid Protector®
Supplier:  LABCONCO
Certificates
undefined
Accessories for storage cabinets for acid Protector®
Supplier:  LABCONCO
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Vent kit
    Vent kit
    LBCP3591100
    3591100
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 3591100
    Local Article Number: LBCP3591100
    Specifications:
    Specifications
    Environmentally Preferable
  • Interior shelf kit with PVC tray for 762 mm protector acid storage cabinets
    Interior shelf kit with PVC tray for 762 mm protector acid storage cabinets
    LBCP9919002
    9919002
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9919002
    Local Article Number: LBCP9919002
    Specifications:
    Specifications
    Environmentally Preferable
  • Interior shelf kit with PVC tray for 914 mm protector acid storage cabinets
    Interior shelf kit with PVC tray for 914 mm protector acid storage cabinets
    LBCP9919001
    9919001
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9919001
    Local Article Number: LBCP9919001
    Specifications:
    Specifications
    Environmentally Preferable
  • Interior shelf kit with PVC tray for 1219 mm protector acid storage cabinets
    Interior shelf kit with PVC tray for 1219 mm protector acid storage cabinets
    LBCP9919000
    9919000
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9919000
    Local Article Number: LBCP9919000
    Specifications:
    Specifications
    Environmentally Preferable
  • Interior shelf kit, 61 mm Protector® acid storage cabinets
    Interior shelf kit, 61 mm Protector® acid storage cabinets
    LBCP9919003
    9919003
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Supplier Article Number: 9919003
    Local Article Number: LBCP9919003
    Specifications:
    Specifications
    Environmentally Preferable