ESD safety shoes, lace-up, Premium CORDURA®
Leverandør: Honeywell Safety Products
Premium Cordura®
DALL93712-517/44EA 1925 NOK
DALL93712-517/44 DALL93712-517/42 DALL93712-517/45 DALL93712-517/48 114-7422 DALL93712-517/38 DALL93712-517/39 DALL93712-517/43 114-7417 114-7416 114-7419 114-7420 114-7423 DALL93712-517/46 114-7414 114-7421 DALL93712-517/41 114-7418 114-7415 DALL93712-517/40 DALL93712-517/49 DALL93712-517/47
ESD safety shoes, lace-up, Premium CORDURA®
Sko og støvler
Sporty, lightweight, ESD safety shoes made from robust, water repellent CORDURA® fabric plus a durable, anti-slip, dual-density TPU/PU sole, resistant to oil and fuel.
  • Premium textile lining
  • PU scuff cap
  • Orthopaedic insole possible
  • Light density, shock-absorbent PU midsole
  • Protective steel toecap
Colour: Black/red
Sertifiseringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.