Glass and stainless steel cleaner, TASKI Sprint Glass
DIVY7513050
: Diversey
- Description:TASKI Sprint Glass, hard surface and glass cleaner
- Packed:6×750 ml
- Pk:6 Bottle
Ammonia-free, glass cleaner to give a non streaking finish on all surfaces including chrome, stainless steel and laminate counter tops.
- Quick drying
- Pleasant fragrance with ONT (Odour Neutralising Technology)