Anti-CD8A Mouse Monoclonal Antibody (CF405M) [clone: C8/144B]

Leverandør: Biotium
BNC050749-100EA 2075 NOK
BNC050749-100 BNC050749-500
Anti-CD8A Mouse Monoclonal Antibody (CF405M) [clone: C8/144B]
Antistoffer
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4 /CD8-. Anaplastic large cell lymphoma is usually CD4 and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: C8/144B
Conjugation: CF405M
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR