These knitted gloves with ¾ dipped neoprene/nitrile coating and backside TPR impact bumpers are ideal for use in the chemical, oil and gas industry.
- Easier handling of tools, increasing safety and reducing fatigue
- Hands stay clean and comfortable when handling wet or oily objects
- Impact protection reduces potential for injury
- 15-gauge liner increases user comfort and dexterity
- High visibility colour increases awareness of location of users' hands for safety
- Intercept™ Technology provides cut protection, increasing safety
Ideal for connection of pipes, disassembly/assembly of motors/tools, handling pipes, bits and other materials, reworking tools, running cables, washing and cleaning tools, heavy machinery/equipment maintenance.
Sertifiseringer: EN 388:2016 (4242BP), EN 407 (X1XXXX); Cat. III