About this item
These cell and tissue culture flasks made of PS are perfect for cell growth.
- Available with five different growth areas
- Flask surface is flat and free from striations to maximise usable growth area
- Two different HDPE cap styles can be used in both open and closed systems
- Innovative angled neck design offers good pipette and cell scraper access
- Triangular top with wide base provides stability
- Easy stacking
- Frosted area near the neck for easy marking/writing
- Engraved graduations on multiple sides
- 100% integrity tested
- Sterilised by electron beam irradiation
- Free from DNase and RNase and non pyrogenic
Non treated: Cell and tissue culture flasks are ideal for applications where cell attachment is not desired.
TC treated: For optimal cell attachment and growth. TC treated, extended (*): Deeper flask for increased recommended working volume.
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype or stressful culture conditions and for those which normally require additional biological coatings for attachment.
734-3419 and 734-3420 perfectly fits for Thermo Fisher incubators. Three can be placed in parallel on each floor to fully use the space.
Specifications
- Catalog No:
- 734-2315
- 734-3253
- 734-2313
- 734-2314
- 734-3252
- 734-2310
- 734-2311
- 734-2312
- 734-2813
- 734-2812
- 734-2792
- 734-2601
- 734-3419
- 734-2791
- 734-3420
- 734-2788
- 734-2810
- 734-2787
- 734-2809
- 734-2790
- 734-2789
- 734-2811
- 734-2784
- 734-2806
- 734-2783
- 734-2805
- 734-2786
- 734-2808
- 734-2785
- 734-2807
- 734-3265
- 734-2316
- 734-2804
- 734-2309
- 734-3266
- 734-2600
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Related Information
Avantor Cell Culture Solutions.pdf VWR Standard Line Products for Tissue Culture VWR® for ovens and incubators
Product Literature
Flasks for suspension cells
Product Information
- Catalog #CapacityGrowth areaSurfaceWorking volumeClosure typeRecommended working volumeAvailabilityPrice
- 734-3253600 ml (*)182,0 cm²Non treated500 mlVented capLog in for availability1 * 40 Itemskr 2 375,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-3252600 ml (*)182,0 cm²Non treated500 mlPlug seal screw capLog in for availability1 * 40 Itemskr 2 150,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-278325 ml12,5 cm²Non-treated20 mlPlug seal screw capLog in for availability1 * 200 Itemskr 3 075,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-278550 ml25,0 cm²Non-treated40 mlPlug seal screw capLog in for availability1 * 200 Itemskr 3 225,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2789600 ml182,0 cm²Non-treated400 mlPlug seal screw capLog in for availability1 * 40 Itemskr 2 300,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2791850 ml300,0 cm²Non-treated800 mlPlug seal screw capLog in for availability1 * 18 Itemskr 2 400,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2787250 ml75,0 cm²Non-treated175 mlPlug seal screw capLog in for availability1 * 100 Itemskr 3 025,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Flasks for adherent cells
Product Information
- Catalog #CapacityGrowth areaSurfaceWorking volumeClosure typeRecommended working volumeAvailabilityPrice
- 734-2315600 ml182,0 cm²TC-treated36,4 – 54,6 mlVented capLog in for availability1 * 40 Itemskr 1 600,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-231025 ml12,5 cm²TC-treated2,5 – 3,75 mlPlug seal screw capLog in for availability1 * 200 Itemskr 3 500,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-230925 ml12,5 cm²TC-treated2,5 – 3,75 mlVented capLog in for availability1 * 200 Itemskr 3 900,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-280425 ml12,5 cm²Increased cell attachment2,5 – 3,75 mlPlug seal screw cap20 mLLog in for availability1 * 200 Itemskr 4 125,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-280525 ml12,5 cm²Increased cell attachment2,5 – 3,75 mlVented cap20 mlLog in for availability1 * 200 Itemskr 4 975,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-231250 ml25,0 cm²TC-treated5 – 7,5 mlPlug seal screw capLog in for availability1 * 200 Itemskr 2 825,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-231150 ml25,0 cm²TC-treated5 – 7,5 mlVented capLog in for availability1 * 200 Itemskr 2 950,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-280650 ml25,0 cm²Increased cell attachment5 – 7,5 mlPlug seal screw cap40 mLLog in for availability1 * 200 Itemskr 4 300,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-280750 ml25,0 cm²Increased cell attachment5 – 7,5 mlVented cap40 mLLog in for availability1 * 200 Itemskr 5 300,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2314250 ml75,0 cm²TC-treated15 – 22,5 mlPlug seal screw capLog in for availability1 * 100 Itemskr 2 425,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2313250 ml75,0 cm²TC-treated15 – 22,5 mlVented capLog in for availability1 * 100 Itemskr 2 425,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2808250 ml75,0 cm²Increased cell attachment15 – 22,5 mlPlug seal screw cap175 mLLog in for availability1 * 100 Itemskr 4 975,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2809250 ml75,0 cm²Increased cell attachment15 – 22,5 mlVented cap175 mLLog in for availability1 * 100 Itemskr 5 750,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-3419400 ml225,0 cm²TC-treated45 - 67,5 mlPlug seal screw capLog in for availability1 * 25 Itemskr 2 800,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-3420400 ml225,0 cm²TC-treated45 - 67,5 mlVented capLog in for availability1 * 25 Itemskr 3 125,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2316600 ml182,0 cm²TC-treated36,4 – 54,6 mlPlug seal screw capLog in for availability1 * 40 Itemskr 1 575,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2810600 ml182,0 cm²Increased cell attachment36,4 – 54,6 mlPlug seal screw cap400 mLLog in for availability1 * 40 Itemskr 3 325,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2601850 ml300,0 cm²TC-treated60 - 90 mlPlug seal screw capLog in for availability1 * 18 Itemskr 1 950,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2812850 ml300,0 cm²Increased cell attachment60 - 90 mlPlug seal screw cap800 mLLog in for availability1 * 18 Itemskr 3 100,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2813850 ml300,0 cm²Increased cell attachment60 - 90 mlVented cap800 mLLog in for availability1 * 18 Itemskr 3 375,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Increased cell attachment surfaceIncreased cell attachment surface734-2811600 ml182,0 cm²Increased cell attachment36,4 – 54,6 mlVented cap400 mLLog in for availability1 * 40 Itemskr 3 975,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-3265600 ml (*)182,0 cm²TC-treated45,5 mlPlug seal screw capLog in for availability1 * 40 Itemskr 1 600,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-3266600 ml (*)182,0 cm²TC-treated45,5 mlVented capLog in for availability1 * 40 Itemskr 1 600,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2600850 ml300,0 cm²TC-treated60 - 90 mlVented capLog in for availability1 * 18 Itemskr 2 125,00Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact