Prolactin
BIRBORB97825-50
: Biorbyt
:
- Pk:50 µG
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 0.065 ng/ml, determined by the dose dependent stimulation of the proliferation of rat lymphoma, Nb2-11.
- UniProtKB:P01240
- Protein/peptide name:Prolactin
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized Prolactinin sterile bidest water not less than 100 g/ml. For longterm storage it is recommended to add a carrier protein(0.1% HSA or BSA).
- Molecular weight:23 kDa
- Formulation:Lyophilized. Prolactin was lyophilized from a solutionwith 0.0045 mM NaHCO3.