Ubbelohde viscometers (DIN) with TC sensors, additional filling, cleaning tube and glass thread
Leverandør: SI Analytics
SCOI285423260EA 16900 NOK
SCOI285423260 SCOI285423140 SCOI285423360 SCOI285423240 SCOI285423340 SCOI285423120 SCOI285423180 SCOI285423280 SCOI285423160 SCOI285423380 SCOI285423110 SCOI285423330 SCOI285423210 SCOI285423310 SCOI285423370 SCOI285423150 SCOI285423250 SCOI285423350 SCOI285423130 SCOI285423230 SCOI285423190 SCOI285423290 SCOI285423170 SCOI285423390 SCOI285423270 SCOI285423400 SCOI285423220 SCOI285423320 SCOI285423200 SCOI285423300
Ubbelohde viscometers (DIN) with TC sensors, additional filling, cleaning tube and glass thread
Especially suitable for liquids that can not be detected with other systems for safe operation.
  • Calibrated with constant for automatic measurement
  • Opaque, black and/or electrically conductive samples
  • Meniscus passage detected due to the different conductivity of the liquid and gas phases
Measuring levels are marked by the TC sensors.
Sertifiseringer: DIN 51562-1, ISO 3105