Heat-on blocks
Blocks
- ForPk
- Heat-On block 250 mlHeat-On block 250 ml442-0636505-80067-0011 * 1 Itemskr 7 250,00
Accessories
Specifications
Environmentally Preferable - Heat-On block 250 ml with side arm cutoutsHeat-On block 250 ml with side arm cutoutsHEID505-80067-01505-80067-0111 * 1 Itemskr 7 700,00
Accessories
Specifications
Environmentally Preferable - Heat-On block 500 mlHeat-On block 500 ml442-0637505-80069-0011 * 1 Itemskr 7 550,00
Accessories
Specifications
Environmentally Preferable - Heat-On block 1 litre
Accessories
Specifications
Environmentally Preferable - Heat-On block 2 litreHeat-On block 2 litre442-0639505-80073-0011 * 1 Itemskr 11 025,00
Accessories
Specifications
Environmentally Preferable - Heat-On block 3 litreHeat-On block 3 litre442-0640505-80075-0011 * 1 Itemskr 13 800,00
Accessories
Specifications
Environmentally Preferable
Heat-On Multi-Well packages
- ForPk
- Heat-On Multi-Well package
including 1× multi-well holder and 6× inserts (2 each for 25, 50 and 100 ml flasks)Heat-On Multi-Well package
including 1× multi-well holder and 6× inserts (2 each for 25, 50 and 100 ml flasks)442-0658505-81200-0011 * 1 Itemskr 17 250,00Accessories
Specifications
Environmentally Preferable
Heat-On PTFE safety covers
- ForPk
- Heat-On Safety cover, PTFEHeat-On Safety cover, PTFEHEID505-80080-00505-80080-00Multi-well holder11 * 1 Itemskr 6 325,00
Accessories
Specifications
Environmentally Preferable - Heat-On Safety cover, PTFEHeat-On Safety cover, PTFEHEID505-80082-00505-80082-00500 ml block11 * 1 Itemskr 7 250,00
Accessories
Specifications
Environmentally Preferable