This fully encapsulated liquid tight chemical suit is designed for use in conjunction with self contained breathing apparatus (SCBA). Developed with the UK Fire & Rescue services.
- Rear entry double zip system
- Rear mounted BA (breathing apparatus) pouch – universal fit with most BA
- Attached socks with boot overflap and attached Ansell Barrier® gloves
- Exhalation valves, clear face visor
- Ultrasonically welded and taped seams
- Bat-wing design enables air gauge checking within the suit
- Chest strap for DSU (Distress Signal Unit), adjustable internal support braces
Typical applications include: Chemicals, oil and petrolchemicals, pharmaceutical, agriculture, sewage purification installations, industrial, tank cleaning and emergency services (HAZMAT, CBRN). Also suitable for use in level-B applications (according to US Environmental Protection Agency (EPA) & NFPA guidelines). Suitable for Ex-Zones.
Sertifiseringer: Type 3/4, EN 14126; Cat. III
Suitable for Ex-Zones