Fully encapsulated suits for use with powered air purifying respirators (PAPR). The lighweight multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film) is designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents. Tested against over 180 chemicals, including chemical warfare agents.
- Double elasticated cuffs: Enables a liquid tight connection with chemical protective glove
- Air permeable SMS collar maintains sufficient air in the breathing zone whilst allowing excess air to flow into the body of the suit
- Attached socks with elasticated boot over-flap
- Panoramic visor design ensures a good field of vision for the wearer
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Four exhalation valves fitted to the rear of the suit allow CO₂ to escape and equalise pressure within the suit
- Utrasonically welded and taped seams provide an effective liquid and particle barrier
For use with Scott Proflow 2SC / EX PAPR fan units, breathing hose and filters.
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
Sertifiseringer: Type 3/4/5, EN 1073-2, EN 14126, EN 1149-1; Cat. III