Fully encapsulated suits for use with powered air purifying respirators (PAPR). The lightweight and durable multi-layer fabric (external film barrier layer with two non woven layers underneath) provides an extremely effective barrier against both inorganic chemicals and biological hazards.
- Double elasticated cuffs: Enables a liquid tight connection with chemical protective glove
- Air permeable SMS collar maintains sufficient air in the breathing zone whilst allowing excess air to flow into the body of the suit
- Attached socks with elasticated boot over-flap
- Panoramic visor design ensures a good field of vision for the wearer
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Four exhalation valves fitted to the rear of the suit allow CO₂ to escape and equalise pressure within the suit
- Utrasonically welded and taped seams provide an effective liquid and particle barrier
For use with Scott Proflow 2SC / EX PAPR fan units, breathing hose and filters.
Typical applications include: Exposure to liquid inorganic chemicals, acids & bases, protection from radioactive particulate contamination.
Sertifiseringer: Type 3/4/5, EN 1073-2, EN 14126, EN 1149-1; Cat. III