Labels for BBP3x printers, type B-7569
Supplier: Brady
Certificates
About this item
B-7569 vinyl is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
- Gloss finish
- Up to three years outdoor use
- Temperature range from –20 to +75 °C
Related Information
Product Information
- Catalog #Supplier #TypeModelMaterialAdhesivePrinting methodAvailabilityPrice
- Label printer tapeLabel printer tapeBRDY120900120900B-7569B30C-500-7569-BKVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120908120908B-7569B30C-500-7569-GNVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120912120912B-7569B30C-500-7569-ORVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120916120916B-7569B30C-500-7569-RDVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120920120920B-7569B30C-500-7569-WTVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120925120925B-7569B30C-500-7569-YLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120901120901B-7569B30C-1125-7569-BKVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120905120905B-7569B30C-1125-7569-BLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120909120909B-7569B30C-1125-7569-GNVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120913120913B-7569B30C-1125-7569-ORVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120917120917B-7569B30C-1125-7569-RDVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120921120921B-7569B30C-1125-7569-WTVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120926120926B-7569B30C-1125-7569-YLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 2 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120902120902B-7569B30C-2250-7569-BKVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120906120906B-7569B30C-2250-7569-BLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120910120910B-7569B30C-2250-7569-GNVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120914120914B-7569B30C-2250-7569-ORVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120918120918B-7569B30C-2250-7569-RDVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120922120922B-7569B30C-2250-7569-WTVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120927120927B-7569B30C-2250-7569-YLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 3 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120923120923B-7569B30C-3000-7569-WTVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120928120928B-7569B30C-3000-7569-YLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 025,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120903120903B-7569B30C-4000-7569-BKVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120904120904B-7569B30C-500-7569-BLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 1 275,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120907120907B-7569B30C-4000-7569-BLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120911120911B-7569B30C-4000-7569-GNVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120915120915B-7569B30C-4000-7569-ORVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120919120919B-7569B30C-4000-7569-RDVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120924120924B-7569B30C-4000-7569-WTVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY120929120929B-7569B30C-4000-7569-YLVinylPermanent acrylicLog in for availability1 * 1 Rullkr 4 525,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120935120935B-7569B30-241-7569-YLVinylPermanent acrylicLog in for availability1 * 300 Itemskr 2 925,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120933120933B-7569B30-219-7569-WTVinylPermanent acrylicLog in for availability1 * 250 Itemskr 2 025,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120930120930B-7569B30-25-7569-WTVinylPermanent acrylicLog in for availability1 * 175 Itemskr 4 525,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120934120934B-7569B30-25-7569-YLVinylPermanent acrylicLog in for availability1 * 175 Itemskr 4 525,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120931120931B-7569B30-241-7569-WTVinylPermanent acrylicLog in for availability1 * 300 Itemskr 2 925,00Specifications:
Specifications
Environmentally Preferable - Pre-cut label tape for printingPre-cut label tape for printingBRDY120932120932B-7569B30-242-7569-WTVinylPermanent acrylicLog in for availability1 * 300 Itemskr 2 025,00Specifications:
Specifications
Environmentally Preferable