Accessories for heating circulators
Supplier: JULABO GmbH
Certificates
About this item
Product Information
- Catalog #Supplier #ForPkAvailabilityPrice
- 2 Adapters M16×1 female to 1/2" tube2 Adapters M16×1 female to 1/2" tubeJULA88900138890013CF, HE2Log in for availability1 * 2 Itemskr 1 150,00Specifications:
Specifications
Environmentally Preferable - Adapter M16×1 male to M16×1 maleAdapter M16×1 male to M16×1 maleJULA89704438970443HE, HL, SL, CF, PLUS, Magnum1Log in for availability1 * 1 Itemskr 520,00Specifications:
Specifications
Environmentally Preferable - 2 Adapters M16x1 male to NPT ¼" female2 Adapters M16x1 male to NPT ¼" femaleJULA88900108890010HE, HL, SL, CF2Log in for availability1 * 2 Itemskr 1 150,00Specifications:
Specifications
Environmentally Preferable - Adapter for metal tubing M10x1 to M16x1Adapter for metal tubing M10x1 to M16x1JULA89704448970444CF30, CF31, CF40, CF411Log in for availability1 * 1 Itemskr 545,00Specifications:
Specifications
Environmentally Preferable - Bath cover with openings and Viton® sleeves I.Ø 2×3 mm, 2×4 mm, 2×6 mmBath cover with openings and Viton® sleeves I.Ø 2×3 mm, 2×4 mm, 2×6 mmJULA897024689702461Log in for availability1 * 1 Itemskr 4 300,00Specifications:
Specifications
Environmentally Preferable - 2 Barbed fittings, M10×1, for 8 mm inner diameter tube2 Barbed fittings, M10×1, for 8 mm inner diameter tubeJULA89704608970460ED, EH, MA, ME2Log in for availability1 * 2 Itemskr 275,00Specifications:
Specifications
Environmentally Preferable - 2 Barbed fittings, M10×1, for 12 mm inner diameter tube2 Barbed fittings, M10×1, for 12 mm inner diameter tubeJULA89704688970468ED, EH, MA, ME2Log in for availability1 * 2 Itemskr 640,00Specifications:
Specifications
Environmentally Preferable - 2 barbed fittings for tubing 12 mm ID2 barbed fittings for tubing 12 mm IDJULA89704458970445CF30, CF31, CF40, CF412Log in for availability1 * 2 Itemskr 905,00Specifications:
Specifications
Environmentally Preferable - D + S level-adapter, to maintain fluid level in an external bathD + S level-adapter, to maintain fluid level in an external bath461-39928970410HE, HL, SE, SL1Log in for availability1 * 1 Itemskr 5 175,00Specifications:
Specifications
Environmentally Preferable - Filling port, insulated with humidity absorberFilling port, insulated with humidity absorberJULA89707058970705SL1Log in for availability1 * 1 Itemskr 6 775,00Specifications:
Specifications
Environmentally Preferable - M + R in-line PT100 sensor with 2× M16×1 male fittingsM + R in-line PT100 sensor with 2× M16×1 male fittings461-39918981020ME, HE, HL, SE, SL1Log in for availability1 * 1 Itemskr 6 450,00Specifications:
Specifications
Environmentally Preferable - MVS solenoid valve controller for tap water coolingMVS solenoid valve controller for tap water cooling461-39939790000MB, MC, ME, HE, SE1Log in for availability1 * 1 Itemskr 9 100,00Specifications:
Specifications
Environmentally Preferable - Platform with castorsPlatform with castorsJULA89100408910040FP40-HE, FP40-HL, FP40-MA, FP40-ME, FP50-HE, FP50-HL, FP50-MA, FPW50-HE, FPW50-HL, FPW50-MA, FPW50-ME1Log in for availability1 * 1 Itemskr 6 275,00Specifications:
Specifications
Environmentally Preferable - Shut off valve for loop circuit, M16×1, −30...+90 °CShut off valve for loop circuit, M16×1, −30...+90 °CJULA89704568970456CF30, CF31, CF40, CF411Log in for availability1 * 1 Itemskr 1 225,00Specifications:
Specifications
Environmentally Preferable - Solenoid valve for tap water cooling, 8 mm Ø tubingSolenoid valve for tap water cooling, 8 mm Ø tubing461-39948980700MB, MC, ME, HE, SE1Log in for availability1 * 1 Itemskr 5 250,00Specifications:
Specifications
Environmentally Preferable - Solenoid valve for tap water cooling, 8 mm Ø tubingSolenoid valve for tap water cooling, 8 mm Ø tubing461-04878980703HL, SL1Log in for availability1 * 1 Itemskr 5 475,00Specifications:
Specifications
Environmentally Preferable - Stainless steel/PTFE coated PT100 sensor with 3 m cable, 300×6 mmStainless steel/PTFE coated PT100 sensor with 3 m cable, 300×6 mmJULA89810158981015CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 7 375,00Specifications:
Specifications
Environmentally Preferable - Stainless steel/PTFE coated PT100 sensor with 3 m cable, 600×6 mmStainless steel/PTFE coated PT100 sensor with 3 m cable, 600×6 mmJULA89810138981013CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 7 475,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 1,5 m cableStainless steel PT100 sensor with 1,5 m cable461-39908981003ME, HE, HL, SE, SL1Log in for availability1 * 1 Itemskr 4 100,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 1,5 m cable, 300×6 mmStainless steel PT100 sensor with 1,5 m cable, 300×6 mm461-39988981010CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 4 375,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 1,5 m cable 20×2 mmStainless steel PT100 sensor with 1,5 m cable 20×2 mm461-39978981006CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 5 775,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 3 m cable, 1200×6 mmStainless steel PT100 sensor with 3 m cable, 1200×6 mmJULA89810148981014CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 10 750,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 3 m cable, 900×6 mmStainless steel PT100 sensor with 3 m cable, 900×6 mmJULA89810168981016CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 9 775,00Specifications:
Specifications
Environmentally Preferable - Stainless steel PT100 sensor with 3 m cable, 200×6 mmStainless steel PT100 sensor with 3 m cable, 200×6 mmJULA89810178981017CF31, CF41, F25-HE, F25-HL, F25-ME1Log in for availability1 * 1 Itemskr 7 025,00Specifications:
Specifications
Environmentally Preferable - M + R in-line PT100 sensor 2×M24×1,5 male fittingsM + R in-line PT100 sensor 2×M24×1,5 male fittings461-14458981021PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A801Log in for availability1 * 1 Itemskr 8 125,00Specifications:
Specifications
Environmentally Preferable - Electronic module with analogue connectionElectronic module with analogue connectionJULA89001008900100FP52-SL, FP52-SL, FP55-SL-150C, FP90-SL, FP90-SL-150C, FPW52-SL, FPW52-SL, FPW55-SL
FPW55-SL-150C, FPW52-SL, FPW52-SL-150C, FPW52-SL, FPW55-SL-150C, FPW90-SL, FPW90-SL, FPW90-SL-150C, FPW91-SL, FPW91-SL1Log in for availability1 * 1 Itemskr 5 800,00Specifications:Specifications
Environmentally Preferable