Norway
Order Entry
Anti-kappa chain (bound) Antibody
  ICNA0855059
Anti-kappa chain (bound) Antibody
  ICNA0855059
 
kr 5 575,00
   
  1 * 2 mL

 

  • Antibody type:
    Secondary
  • Antigen name:
    Kappa Chain (bound)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antibody fragment:
    F(ab´)2
  • Storage buffer:
    pH 7.3
  • Concentration:
    Total Protein: 20.4 mg/mL; Antibody Protein: 8 mg/mL
  • Shipping temperature:
    RT
  • Purification:
    The F(ab')2 fragment is prepared from the specific goat IgG fraction by limited digestion with pepsin, and purified by gel filtration or affinity chromatography. The resulting F(ab')2 fragment is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 μm filter, vialed and lyophilized.
  • Size:
    2 ml
  • Pk:
    2 mL

 

 

The total protein is measured using the Biuret procedure, with bovine albumin as a standard. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. F(ab')2 fragments are tested for purity and specificity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is goat F(ab')2; no trace of albumin, whole molecule IgG or Fc is detected.

Type: Secondary
Antigen: Kappa Chain (bound)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human