Yttrium plasma standard solutions, BAKER INSTRA-ANALYZED®, J.T.Baker®
BAKR5755.0100EA 3200 NOK
BAKR5755.0100 BAKR5790.0100
Yttrium plasma standard solutions, BAKER INSTRA-ANALYZED®, J.T.Baker®
Single element plasma standards are used for accuracy and easy of calibration, along with precise detection of trace impurities.
Sertifiseringer: Traceable to NIST.
NB: 10,000 µg/ml= (1.00% w/v)= 10000 mg/l
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022