Size exclusion and ligand exchange chromatography columns, SUGAR KS-803 to KS-807
Supplier: Resonac
Certificates
About this item
These columns separate oligo- and polysaccharides by size exclusion mode.
- Only water is required for the analysis of neutral sugars
- Can be used in combination with other KS series columns
- Base material is styrene divinylbenzene copolymer
* Estimated value
Specifications
- Catalog No:
- 554-2217
- 554-2216
- 554-2227
- Description:
- SUGAR KS-804
- SUGAR KS-803
- SUGAR KS-G 6B
- Max. pressure:
- 5,0 MPa (50 bar)
- 5,0 MPa (50 bar)
- pH range:
- 3 - 7
- 3 - 7
- 3 - 7
- Temperature limit:
- 20 - 85 °C
- 20 - 85 °C
- 20 - 85 °C
- USP code:
- L22, L58
- L22, L58
- L22, L58
Specifications
Product Information
- Catalog #Supplier #Column FormatColumn hardwareParticle SizeØ int.LengthAvailabilityPrice
- SUGAR KS-803SUGAR KS-803554-2216F6378025AnalyticalStainless steel6 µm8,00 mm300 mmLog in for availability1 * 1 Itemskr 23 075,00Specifications:
More
Specifications
DescriptionSUGAR KS-803Environmentally PreferableMax. pressure5,0 MPa (50 bar)pH range3 - 7Temperature limit20 - 85 °CUSP codeL22, L58 - SUGAR KS-804SUGAR KS-804554-2217F6378035AnalyticalStainless steel7 µm8,00 mm300 mmLog in for availability1 * 1 Itemskr 23 075,00Specifications:
More
Specifications
DescriptionSUGAR KS-804Environmentally PreferableMax. pressure5,0 MPa (50 bar)pH range3 - 7Temperature limit20 - 85 °CUSP codeL22, L58
Guard columns
Product Information
- Catalog #Supplier #Column FormatColumn hardwareParticle SizeØ int.LengthAvailabilityPrice
- SUGAR KS-G 6BSUGAR KS-G 6B554-2227F6700020Guard columnStainless steel10 µm6,00 mm50 mmLog in for availability1 * 1 Itemskr 7 600,00Specifications:
More
Specifications
DescriptionSUGAR KS-G 6BEnvironmentally PreferableMax. pressurepH range3 - 7Temperature limit20 - 85 °CUSP codeL22, L58