Anti-SLC9A1 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BOSSBS-0505R-CY7EA 9875 NOK
BOSSBS-0505R-CY7
Anti-SLC9A1 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Plays an important role in signal transduction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC9A1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR