- Antibody type:Primary
- Antigen name:Synaptic Ras GTPase-activating protein 1
- Antigen symbol:SYNGAP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8831
- Antigen synonyms:ras GTPase-activating protein SynGAP|SYNGAP1|MRD5|DKFZp761G1421|synaptic Ras GTPase activating protein 1 homolog|synaptic Ras GTPase activating protein 1|KIAA1938|neuronal RasGAP|RASA5|RASA1
- Modification:phospho-Ser836
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The PSD-95/SAP 90 family of proteins, which are known to bind to and cluster various membrane proteins, are involved in the organization of synaptic structure. SynGAP (for synaptic GTPase activating protein), a novel synaptic RasGAP, is a brain-specific protein abundant in the cortex, hippocampus and olfactory bulb. SynGAP interacts with all three PDZ domains within PSD-95/ SAP 90 proteins. It represents one of three classes of mammalian RasGAPs and is specifically localized to excitatory synapses possessing NMDA receptors. SynGAP may be involved in the regulation of BDNF as well as Ras signaling. Its activity is inhibited by phosphorylation by CaMKII, which could result in the activation of the MAP kinase pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SYNGAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat