Norway
Order Entry
Anti-ELMO1 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-ELMO1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5807R-CY5.5
Supplier:  Bioss
Anti-ELMO1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5807R-CY5.5
Supplier:  Bioss
Supplier Article Number: BS-5807R-CY5.5
Local Article Number: BOSSBS-5807R-CY5.5
Price
kr 10 550,00
Log in for availability
Unit of Measure 1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    engulfment and cell motility 1
  • Antigen symbol:
    ELMO1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9844
  • Antigen synonyms:
    MGC126406|ced-12 homolog 1|ELMO-1|engulfment and cell motility 1|KIAA0281|CED12|CED-12
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

ELMO1 is involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. ELMO1 acts in association with DOCK1 and CRK and was initially proposed to be required in complex with DOCK1 to activate Rac Rho small GTPases. It is belived to enhance the guanine nucleotide exchange factor (GEF) activity of DOCK1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat